Learn More
Research Products International Corp Micro-Centrifuge Tubes, 1.5ml Capacity, Graduated 100ul, Non-Stick, Green, 500 per Case
SDP
Catalog No. 50998586
Encompass
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.
Catalog No. 50-998-586 Supplier Research Products International Corp Supplier No. 145530G
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.