Learn More
Sarstedt Inc MICRO TUBES 2.0ML, TYPE I
Supplier:  Sarstedt Inc 72.694.217
Encompass_Preferred
Micro tubes 2 ml, PP, with knurls, with skirted base, with attached unscrewed screw cap, Biosphere certified (sterile, DNA, RNase, and ATP-free and non-pyrogenic). 25 pieces/bag, 250pc inner bag u 2 bags per case (500 tubes total).
Catalog No. NC0443122