Learn More
Research Products International Corp UltraFlux Strip Tubes with Unattached Caps, Natural Color, 0.2ml Capacity, 8 Tubes per Strip, 120 Strips per Case
SDP
Catalog No. 50212315
Encompass
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. Supplied as 125 strips of 8 per case. Total of 1,000 tubes.
Catalog No. 50-212-315 Supplier Research Products International Corp Supplier No. 145570
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. Supplied as 125 strips of 8 per case. Total of 1,000 tubes.