Learn More
Research Products International Corp UltraFlux Strip Tubes with Unattached Caps, Assorted Colors, 0.2ml Capacity, 8 Tubes per Strip, 120 Strips per Case
SDP
Catalog No. 50212316
Encompass
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation.
Catalog No. 50-212-316 Supplier Research Products International Corp Supplier No. 145578
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation.