Learn More
Research Products International Corp UltraFlux SnapStrip Micro-Tubes, 0.2ml Capacity, Assorted Colors, 120 Strips per Case
SDP
Catalog No. 50212320
Encompass
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. 960 tubes per case.
Catalog No. 50-212-320 Supplier Research Products International Corp Supplier No. 145598
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. 960 tubes per case.