Learn More
Research Products International Corp UltraFlux 12 Strip Tubes with Unattached Cap Strip, Assorted Colors, 0.2ml Capacity, 80 Strips per Case
SDP
Catalog No. 50212318
Encompass
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation.
Catalog No. 50-212-318 Supplier Research Products International Corp Supplier No. 145588
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation.