Learn More
Research Products International Corp Micro-Tubes, Graduated, 0.5ml, Red, 1000/cs
SDP
Catalog No. 501978287
Encompass
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C.
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-197-8287 Supplier Research Products International Corp Supplier No. 145505R
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C.
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.