Learn More
Research Products International Corp Micro-Tubes, Graduated, 500ul, Red, 100/cs
SDP
Catalog No. 501978706
Encompass
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C. 
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-197-8706 Supplier Research Products International Corp Supplier No. 145506R
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C. 
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.