Learn More
Roboz Surgical MICRO VASCULAR CLIP 11MM
SDP
Catalog No. NC9758998
Encompass
Micro Vascular Clip; 1-2mm Vessel Diameter; 35-40 Grams Occlusion Pressure; 11mm Length Please use the RS-6496 applying forcepts with these clips. Compressing the clips with fingers is very difficult and it may damage them. Not for human use
Catalog No. NC9758998 Supplier Roboz Surgical Supplier No. RS6472
Micro Vascular Clip; 1-2mm Vessel Diameter; 35-40 Grams Occlusion Pressure; 11mm Length Please use the RS-6496 applying forcepts with these clips. Compressing the clips with fingers is very difficult and it may damage them. Not for human use