Learn More
Molecular Devices Molecular Devices Fatty Acid Uptake Assay Kit, Homogeneous, Fluorescence, 96/384-Well
Catalog No. 5000014368
Encompass_Preferred
The QBT Fatty Acid Uptake Assay Kit is a homogeneous, fluorescence-based assay for rapid, real-time measurement of fatty acid uptake in live cells. It uses a BODIPY-dodecanoic acid fluorescent analog with quench technology and is configured for high-throughput screening in 96- and 384-well microplates.
  • One-step homogeneous fluorescence assay using a BODIPY-dodecanoic acid analog.
  • Proprietary quench-based format for real-time fatty acid uptake monitoring.
  • Evaluation kit contains 2 vials, sufficient for two microplates.
  • Compatible with 96-well and 384-well microplate formats and bottom-read detection.
  • Designed for high-throughput screening of fatty acid transporters and uptake inhibitors.
Catalog No. 50-000-14368 Supplier Molecular Devices Supplier No. R6132
The QBT Fatty Acid Uptake Assay Kit is a homogeneous, fluorescence-based assay for rapid, real-time measurement of fatty acid uptake in live cells. It uses a BODIPY-dodecanoic acid fluorescent analog with quench technology and is configured for high-throughput screening in 96- and 384-well microplates.
  • One-step homogeneous fluorescence assay using a BODIPY-dodecanoic acid analog.
  • Proprietary quench-based format for real-time fatty acid uptake monitoring.
  • Evaluation kit contains 2 vials, sufficient for two microplates.
  • Compatible with 96-well and 384-well microplate formats and bottom-read detection.
  • Designed for high-throughput screening of fatty acid transporters and uptake inhibitors.