Learn More
Andwin Scientific MICRO WEIGH DISH .35ML 100/PK
SDP
Catalog No. 501305909
Encompass_Preferred
The 13mm disposable micro aluminum weighing dishes are ideal for microbalances, organic micro elementary, thermal analysis and pharmaceutical use. It has a capacity of 0.23ml and is sold as a pack of 100.
Catalog No. 50-130-5909 Supplier Andwin Scientific Supplier No. 101907A
The 13mm disposable micro aluminum weighing dishes are ideal for microbalances, organic micro elementary, thermal analysis and pharmaceutical use. It has a capacity of 0.23ml and is sold as a pack of 100.