Learn More
Andwin Scientific Aluminum Micro Weighing Dishes, 12mm W x 2mm L x 4.5mm H
SDP
Catalog No. NC0058257
Encompass_Preferred
Aluminum Micro Weighing Dishes These micro weigh boats are compatible with most commercially available analyzers and micro balances. Capacity: 0.12mL. Size: 12mm W x 2mm L x 4.5mm H.
Catalog No. NC0058257 Supplier Andwin Scientific Supplier No. 12577072
Aluminum Micro Weighing Dishes These micro weigh boats are compatible with most commercially available analyzers and micro balances. Capacity: 0.12mL. Size: 12mm W x 2mm L x 4.5mm H.