Learn More
Premier Lab Supply Micro XRF Sample Cups – PN# MC-1520
SDP
Catalog No. NC2057165
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
31mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# MC-1520) – OD: 31mm; ID: 17mm; Height: 21mm; Volume: 4mL – 100 per bag
Catalog No. NC2057165 Supplier Premier Lab Supply Supplier No. MC1520
This item has a minimum qty of 2 per supplier requirements.
31mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# MC-1520) – OD: 31mm; ID: 17mm; Height: 21mm; Volume: 4mL – 100 per bag