Learn More
Lonza Walkersville MycoZap Plus-CL
Catalog No. NC0214412
Encompass
MycoZap Plus-CL is a ready-to-use solution of innovative antibiotics for protecting continuous cell lines cultures from a broad range of common contaminants. MycoZap Plus-PR is active against mycoplasma, but also Gram(-) and Gram(+) bacteria as well as yeast and fungi. MycoZap Plus-PR can be used continuously and replace the old-fashioned Pen/Strep formulation.Benefits:Active against mycoplasma, Gram(-) and Gram(+) bacteria as well as yeast and fungiComplete solution replacing Pen/Strep formulationApplications:MycoZap Plus-CL for protection of cell linesContent:10 x 1 mL MycoZap Plus-CL Antibiotic
Catalog No. NC0214412 Supplier Lonza Walkersville Supplier No. VZA2011
MycoZap Plus-CL is a ready-to-use solution of innovative antibiotics for protecting continuous cell lines cultures from a broad range of common contaminants. MycoZap Plus-PR is active against mycoplasma, but also Gram(-) and Gram(+) bacteria as well as yeast and fungi. MycoZap Plus-PR can be used continuously and replace the old-fashioned Pen/Strep formulation.Benefits:Active against mycoplasma, Gram(-) and Gram(+) bacteria as well as yeast and fungiComplete solution replacing Pen/Strep formulationApplications:MycoZap Plus-CL for protection of cell linesContent:10 x 1 mL MycoZap Plus-CL Antibiotic