Learn More
Sino Biological Peptide Substrates: Micro2 Peptide
Catalog No. 502568695
Encompass_Preferred
The Micro2 Peptide sequence (KEEQSQITSQVTGQIGWR) is derived from human AP-2 complex subunit mu isoform b (amino acids 143-160) and is suitable as a substrate for AAK1, BMP2K, and GAK kinase assay.
Catalog No. 50-256-8695 Supplier Sino Biological Supplier No. M08581000
The Micro2 Peptide sequence (KEEQSQITSQVTGQIGWR) is derived from human AP-2 complex subunit mu isoform b (amino acids 143-160) and is suitable as a substrate for AAK1, BMP2K, and GAK kinase assay.