Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories MicroAlbumin 2-1 Combo Strips

Catalog No. 23111312
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-312 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-312 Supplier Germaine™ Laboratories Supplier No. 52255
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories MicroAlbumin 2-1 Combo Dipstick reagent strips are for the qualitative detection of Microalbumin and Creatinine

  • Urine dipstick for Microalbumin and Creatinine
  • Determines Albumin to Creatinine ratio, CLIA Waived
  • For visual read only, analyzer not required

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Microalbumin, Creatinine
Sample Type Urine
Disposable Single-use
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.