Learn More
Cliawaived Inc Microalbumin 2-1 Combo Strips
SDP
Catalog No. NC1254280
Encompass_Preferred
(25/Bottle) The Combo Strip is designed to give semi-quantitative results for Microalbumin and Creatinine in random urine and also determine the albumin to creatinine ratio in random urine. The Microalbumin 2-1 combo Strip is a plastic strip to which two test pads are affixed testing for Microalbumin and Creatinine. Microalbuminuria, an abnormal elevation of the urinary albumin excretion rate, is many times one of the first signs of renal diseases and renal damage that can lead to renal failure. Patients with the highest risk of renal disease where Microalbumin may be present is hypertension and diabetes.
This item is not returnable. View return policy
Catalog No. NC1254280 Supplier Cliawaived Inc Supplier No. TCURS2M
(25/Bottle) The Combo Strip is designed to give semi-quantitative results for Microalbumin and Creatinine in random urine and also determine the albumin to creatinine ratio in random urine. The Microalbumin 2-1 combo Strip is a plastic strip to which two test pads are affixed testing for Microalbumin and Creatinine. Microalbuminuria, an abnormal elevation of the urinary albumin excretion rate, is many times one of the first signs of renal diseases and renal damage that can lead to renal failure. Patients with the highest risk of renal disease where Microalbumin may be present is hypertension and diabetes.