Learn More
Fujifilm Healthcare Americas Corp Autokit Microalbumin Calibrator Set
Catalog No. NC1922876
Encompass_Preferred
The Autokit Microalbumin Calibrator Set, which contains 5 calibrators with different albumin concentration, is designed to be used with Autokit Micro Albumin test for the determination of urinary albumin. Package Size: Calibrator 1 1 bottle x for 2 mL, Calibrator 2 1 bottle x for 2 mL, Calibrator 3 1 bottle x for 2 mL, Calibrator 4 1 bottle x for 2 mL, Calibrator 5 1 bottle x for 2 mL. This product is For In Vitro Diagnostic Use. Custom order: Contact supplier for additional information at www.ivd.fujimed.com.
This item is not returnable. View return policy
Catalog No. NC1922876 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99506101
The Autokit Microalbumin Calibrator Set, which contains 5 calibrators with different albumin concentration, is designed to be used with Autokit Micro Albumin test for the determination of urinary albumin. Package Size: Calibrator 1 1 bottle x for 2 mL, Calibrator 2 1 bottle x for 2 mL, Calibrator 3 1 bottle x for 2 mL, Calibrator 4 1 bottle x for 2 mL, Calibrator 5 1 bottle x for 2 mL. This product is For In Vitro Diagnostic Use. Custom order: Contact supplier for additional information at www.ivd.fujimed.com.