Learn More
Fujifilm Healthcare Americas Corp Autokit Microalbumin Control Set
Catalog No. NC1922877
Encompass_Preferred
The Autokit Microalbumin Control Set, which contains 2 controls with different albumin concentration , is designed to be used with the Autokit Microalbumin assay for the determination of urinary albumin. Package size: Microalbumin Control 1 2 bottles x 2 mL and Microalbumin Control 2 2 bottles x 2 mL. This product is For In Vitro Diagnostic Use. Custom order: Contact supplier for additional information at www.ivd.fujimed.com.
This item is not returnable. View return policy
Catalog No. NC1922877 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99106201
The Autokit Microalbumin Control Set, which contains 2 controls with different albumin concentration , is designed to be used with the Autokit Microalbumin assay for the determination of urinary albumin. Package size: Microalbumin Control 1 2 bottles x 2 mL and Microalbumin Control 2 2 bottles x 2 mL. This product is For In Vitro Diagnostic Use. Custom order: Contact supplier for additional information at www.ivd.fujimed.com.