Learn More
Quantimetrix Microbumin Microalbumin Control Set Level 1&2
SDP
Catalog No. NC9733589
Encompass_Preferred
3-year refrigerated shelf life from date of manufacture6-month open vial stability when stored at 2C8CLiquid, made with human urineIncludes creatinineWith longer stability and less waste, Microbumin Microalbumin Control monitors the performance of popular strips including Clinitek Microalbumin Reagent Strips and Micral™ test strips. Microbumin is also perfect for use with turbidimetric immunoassays using Siemens DCA 2000, Siemens Dimension, Beckman Coulter Synchron, Beckman Coulter AU Instruments, Abbott Architect, Roche cobas and nephelometry.
Catalog No. NC9733589 Supplier Quantimetrix Supplier No. 125001
3-year refrigerated shelf life from date of manufacture6-month open vial stability when stored at 2C8CLiquid, made with human urineIncludes creatinineWith longer stability and less waste, Microbumin Microalbumin Control monitors the performance of popular strips including Clinitek Microalbumin Reagent Strips and Micral™ test strips. Microbumin is also perfect for use with turbidimetric immunoassays using Siemens DCA 2000, Siemens Dimension, Beckman Coulter Synchron, Beckman Coulter AU Instruments, Abbott Architect, Roche cobas and nephelometry.