Learn More
Kamiya Biomedical Microalbumin Urine Control, L1: 2 x 7mL, L2: 2 x 7mL RX
SDP
Catalog No. 501305929 Shop All Kamiya Biomedical Products
Encompass_Preferred
K-ASSAY Microalbumin Urine Control. Level 1: 2 x 7 mL (liquid), Level 2: 2 x 7 mL (liquid). The K-ASSAY Microalbumin Urine Control is intended for use as an assayed quality control material for monitoring the performance of the K-ASSAY Microalbumin immunoturbidimetric assay (Cat. No. KAI-019 or KAI-057). FOR IN VITRO DIAGNOSTIC USE.
Catalog No. 50-130-5929 Supplier Kamiya Biomedical Supplier No. K37C4M
K-ASSAY Microalbumin Urine Control. Level 1: 2 x 7 mL (liquid), Level 2: 2 x 7 mL (liquid). The K-ASSAY Microalbumin Urine Control is intended for use as an assayed quality control material for monitoring the performance of the K-ASSAY Microalbumin immunoturbidimetric assay (Cat. No. KAI-019 or KAI-057). FOR IN VITRO DIAGNOSTIC USE.