Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 3 x 32-Well Retainers

Catalog No. 4481669
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 retainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-816-69 1 retainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-816-69 Supplier Applied Biosystems™ Supplier No. 4481669
Add to Cart
Add to Cart

Designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System

  • The Applied Biosystems MicroAmp 3 x 32-Well Retainer is specifically designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System.
  • The retainer provides support and allows pressure to be distributed evenly over tubes and strips to ensure a proper seal and prevent the tubes and strips from being crushed or deformed.
  • Its use is recommended at all times when working with MicroAmp reaction tubes and tube strips.
  • Simply place the retainer on the sample block, then load the tubes or strips.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.

Product Line MicroAmp
Color Black
Product Type Retainer
For Use With (Equipment) ProFlex 3 x 32-Well PCR System
For Use With (Application) PCR
Plate Compatibility 32-Well Plate
Quantity 1 retainer
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.