Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog No. 403083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Assemblies
20 Assemblies
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-308-3 20 Assemblies
40-308-6 100 Assemblies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-308-3 Supplier Applied Biosystems™ Supplier No. 403083
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

20 MicroAmp 96-Well Reaction Tube/Tray Retainer Sets.

Designed for precise PCR on Applied Biosystems 0.2mL thermal cyclers

Set contains MicroAmpreaction tubes without caps already loaded in a MicroAmp 96-Well Tray/Retainer.

  • Completely pre-assembled for convenience
  • Available in economical 100-unit bulk packs
  • Non-sterile and autoclavable
  • Greater Sample Uniformity
  • Use with all instruments that have heated sample covers to eliminate condensation and need for mineral oil overlay
  • Proprietary MicroAmpreaction tubes are essential for high-performance PCR and ensure greatest temperature and amplification uniformity for samples
  • MicroAmp 96-Well Reaction Tube/Tray/Retainer Set requires caps or full plate covers to seal tubes

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 20 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.
Product Line MicroAmp
Color Natural
Product Type 96-Well Reaction Tube/Tray/Retainer Set
For Use With (Equipment) 2720 Thermal Cycler, 7000 System, AB 2720 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 20 Assemblies
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.