Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate

Catalog No. n8010560
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
500 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010560 10 Plates
43-168-13 500 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N8010560 Supplier Applied Biosystems™ Supplier No. N8010560
  • $93.00
    $87.65 / Each of 1

    Save $5.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems 96-well Real-Time PCR systems and thermal cyclers.

  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Capacity: 96 x 0.2mL tubes

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 6100 Nucleic Acid PrepStation, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

Applications: PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains ten 96-well reaction plates.

Will require either caps or full plate cover to seal (not included).

May be stored at room temperature. With proper storage, plates may be stored indefinitely.
Packaging 10 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 10 Plates
Shipping Condition Room Temperature
Show More Show Less
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?

Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.

Which instruments is the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4416813) compatble with?

The MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4416813) is constructed from a single rigid piece of polypropylene and is compatible with Applied Biosystems 96-well Real-Time PCR systems and thermal cyclers.

Is the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4416813) DNA/RNAse-free?

Yes, the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4416813) is certified DNA/RNAse-free.

Are the MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737, 4326659) and the one without Barcode (Cat. No. N8010560, 4316813) the same product?

Yes, both the plates are the same. The only difference between them is the barcode.

Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?

No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.

Is the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) compatible with the SeqStudio Genetic Analyzer?

Yes, the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) is compatible with the SeqStudo Genetic Analyzer.

Is the 96 well plate in SequalPrep Normalization Plate Kit (Cat. No. A1051001) the same as the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813)?

No, they are not the same. The MicroAmp Optical 96-Well Reaction Plate is an empty optical plate that is used for sample processing (i.e. amplification using the thermal cycler or sample loading onto a capillary instrument). The MicroAmp plate cannot be used for performing normalization as it is not coated using the ChargeSwitch technology.

What is the difference between the MicroAmp Optical Adhesive Covers and the MicroAmp Optical Caps? Can they be used interchangeably?

The MicroAmp Optical Adhesive Covers (P/N 4311971) have a higher optical efficiency than the MicroAmp Optical Caps, (8 caps/strip) (P/N 4323032). They can be used instead of the MicroAmp Optical Caps only when using the MicroAmp 96-Well Reaction Plate with Barcode (P/N 4306737, 4309849) or the MicroAmp Optical 96-Well Reaction Plate (P/N N801-0560). The Optical Adhesive Covers cannot be used with the MicroAmp Optical Tubes.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.