Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Base

Catalog No. n8010531
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010531 10 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N8010531 Supplier Applied Biosystems™ Supplier No. N8010531
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 MicroAmp 96-Well Bases.

Convenient for pre- and post-PCR sample processing

Applied Biosystems MicroAmp Bases are designed to hold MicroAmp Reaction Tubes in Tray/Retainer Sets or MicroAmp 96-Well Reaction Plates for convenient use in pre- and post-PCR sample processing.

• Convenient for pre- and post-PCR sample processing
• Easy to use for pipetting steps
• Available in 96-well plate format

Uniform Results
Use with all instruments that have heated sample covers, to eliminate condensation and the need for a mineral oil overlay. Proprietary MicroAmp Reaction Tubes are essential for high-performance PCR. They ensure the greatest temperature and amplification uniformity for your samples.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ 96-Well Bases. Store at room temperature.
Product Line MicroAmp
Color Black
Product Type 96-Well Base
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 10 Pieces
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.