Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates

Catalog No. A36931
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36931 20 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36931 Supplier Applied Biosystems™ Supplier No. A36931
  • $212.00
    $201.65 / Each of 1

    Save $10.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs.

EnduraPlate™ plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. They are constructed to ANSI/SBS standards and are well-suited for automated workflows. MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates are skirted and fit 384-well (0.02 mL) thermal blocks.

  • Optimized for performance with Applied Biosystems™ instruments
  • Certified DNA-, RNase-, and PCR inhibitor-free
  • Constructed to ANSI/SBS standards for compatibility with high-throughput laboratories and robotic handling
  • Easy-to-read black well-identification text to minimize pipetting errors

EnduraPlate™ reaction plates use polypropylene for a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from the thermal block to the sample, while a polycarbonate frame provides mechanical stability, flatness, and durability that is compatible with automation/robotics. Easy-to-read, black well-identification text helps to minimize pipetting mistakes and improves sample preparation.

MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates are provided without barcodes, enabling customization with your own barcodes.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Optical, Clear
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Skirted
Material Polycarbonate, Polypropylene
No. of Wells 384
Quantity 20 plates
Shipping Condition Room Temperature
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.