Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 96-Well GPLE Reaction Plates

Catalog No. 4481191
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-811-91 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-811-91 Supplier Applied Biosystems™ Supplier No. 4481191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heat transfer from thermal block to sample with thin-wall polypropylene wells

  • Applied Biosystems MicroAmp Optical 96-Well GPLE Reaction Plates are validated and tested for optimal fit and performance on Applied Biosystems standard (0.2 mL) 96-well thermal cyclers, 0.2-mL real-time PCR instruments, and genetic analyzers.
  • Features of the MicroAmp Optical 96-Well GPLE Reaction Plates include
  • Each lot is certified DNA-, RNase-, and PCR inhibitor-free
  • Labeled “For Laboratory Use”
  • Optimized for performance with Applied Biosystems instruments
  • Excellent heat transfer from thermal block to sample with thin-wall polypropylene wells
  • Raised deck for efficient plate sealing to help prevent evaporation and minimize contamination
  • MicroAmp Optical 96-well reaction plates are constructed from a single rigid piece of polypropylene and include a raised deck for efficient plate sealing. The thin-wall polypropylene wells provide excellent heat transfer from the thermal block to the sample for fast, efficient PCR amplification.
  • With General Purpose Lab Equipment (GPLE) plastics, each lot of reaction plates is certified in an ISO 13485-registered facility to be free of DNA, RNase, and PCR inhibitors. GPLE plastics are ideal for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Packaging 10 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 10 Plates
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.