Learn More
Electron Microscopy Sciences MAC Microanalysis Standard Manganese Grain, 2 x 2 mm
SDP
Catalog No. 5026350
Encompass_Preferred
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Grain of standard measures approximately 2mm x 2mm in size.
Type: Grain
Size: 2 x 2 mm
Catalog No. 50-263-50 Supplier Electron Microscopy Sciences Supplier No. 80075MN
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Grain of standard measures approximately 2mm x 2mm in size.
Type: Grain
Size: 2 x 2 mm