Learn More
ATCC Clostridioides difficile
Supplier:  ATCC BAA1805
Encompass
This strain of Clostridioides difficile is a clinical isolate with applications in enteric disease research. This strain is toxinotype IIIB and is positive for the presence of the cdtB, tcdA, and tcdB genes.
Catalog No. 50-238-0756