Learn More
Bioworld Replica Plating Tool (100 mm)
Catalog No. NC0400773
Encompass_Preferred
For replicating 100mm plates. Microbe replica platers for efficient and rapid transfer of bacterial and yeast cultures to a fresh plate. Reusable velvet or velveteen squares are used to cover the upper surface of the replica plater and act as the substrate for accurate transfers. Solid metal ring with lock nut slips around the circumference of the plate contact surface and holds valvet squares firmly in place during use. Solid metal construction will last virtually forever. Use with sterilized 6 x 6 inch Velvet Pads (43170019-1) to transfer colonies while retaining colony orientation.
Catalog No. NC0400773 Supplier Bioworld Supplier No. 431700041
For replicating 100mm plates. Microbe replica platers for efficient and rapid transfer of bacterial and yeast cultures to a fresh plate. Reusable velvet or velveteen squares are used to cover the upper surface of the replica plater and act as the substrate for accurate transfers. Solid metal ring with lock nut slips around the circumference of the plate contact surface and holds valvet squares firmly in place during use. Solid metal construction will last virtually forever. Use with sterilized 6 x 6 inch Velvet Pads (43170019-1) to transfer colonies while retaining colony orientation.