Learn More
Sigma Aldrich Fine Chemicals Biosciences MicroBead Sterilizer with1EA
Catalog No. 502908905
Encompass
Benchmarks Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps scissors etc.). When the chamber is filled with the included glass beads high temperatures (up to 300C) can be used to eliminate the presence of bacteria spores and other microorganisms.Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas flame or chemicals and is safe for use in hoods.With adjustable temperature the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools.This product is designed for RESEARCH USE ONLY.Not intended for clinical use.
Catalog No. 50-290-8905 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7425571EA
Benchmarks Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps scissors etc.). When the chamber is filled with the included glass beads high temperatures (up to 300C) can be used to eliminate the presence of bacteria spores and other microorganisms.Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas flame or chemicals and is safe for use in hoods.With adjustable temperature the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools.This product is designed for RESEARCH USE ONLY.Not intended for clinical use.