Learn More
Microsolv Technology Corporation Microbee Silica 4.6 X250Mm Std
Catalog No. 5001451163
Encompass_Preferred
Cogent NP Silica Microbee Similars™ 10µm, 125Å: Compares to Microbondapak™ Silica, 250 x 4.6mm.
Catalog No. 50-014-51163 Supplier Microsolv Technology Corporation Supplier No. 5900125P
Cogent NP Silica Microbee Similars™ 10µm, 125Å: Compares to Microbondapak™ Silica, 250 x 4.6mm.