Learn More
PCE AMERICAS INC PCE-AS1 portable air sampling meter used to assess microbiological pollution and monitor hygiene
SDP
Catalog No. NC3654617
Encompass_Preferred
"Portable 100 lpm aerosol hygiene monitoring device for airborne microbial, microbiological, bacterial, mold, fungi and virus analysisPCE-AS1 is an air sampling meter used to assess microbiological pollution and monitor hygiene. Designed for use in hospitals, clean rooms and food and beverage, cosmetics and pharmaceutical production facilities, the PCE-AS1 air sampler meets the latest requirements for devices used for airborne microbial analysis. Optional ISO calibration lab certificate as an accessory."
Catalog No. NC3654617 Supplier PCE AMERICAS INC Supplier No. PCEAS1
"Portable 100 lpm aerosol hygiene monitoring device for airborne microbial, microbiological, bacterial, mold, fungi and virus analysisPCE-AS1 is an air sampling meter used to assess microbiological pollution and monitor hygiene. Designed for use in hospitals, clean rooms and food and beverage, cosmetics and pharmaceutical production facilities, the PCE-AS1 air sampler meets the latest requirements for devices used for airborne microbial analysis. Optional ISO calibration lab certificate as an accessory."