Learn More
Zymo Research Corporation ZymoBIOMICS™ Microbial Community DNA Standard (2000 ng)
SDP
Catalog No. NC1244170
Encompass_Preferred
One of the major challenges in the emerging field of microbiomics is the bias and errors introduced in the complex workflows. Besides nucleic acid purification, bias also arises from sequencing library preparation and subsequent processes. The ZymoBIOMICS Microbial Community DNA Standard is designed to assess bias, errors and other artifacts after the step of nucleic acid purification. This DNA standard is created by pooling DNA extracted from pure cultures. It has an accurately defined composition, negligible impurities (0.01%), and contains genomes of a wide range of GC content (15% - 85%). This DNA standard is designed to have the same microbial composition with the cellular version, the ZymoBIOMICS Microbial Community Standard, so that they can be more powerful when working in tandem.
Catalog No. NC1244170 Supplier Zymo Research Corporation Supplier No. D6306
One of the major challenges in the emerging field of microbiomics is the bias and errors introduced in the complex workflows. Besides nucleic acid purification, bias also arises from sequencing library preparation and subsequent processes. The ZymoBIOMICS Microbial Community DNA Standard is designed to assess bias, errors and other artifacts after the step of nucleic acid purification. This DNA standard is created by pooling DNA extracted from pure cultures. It has an accurately defined composition, negligible impurities (0.01%), and contains genomes of a wide range of GC content (15% - 85%). This DNA standard is designed to have the same microbial composition with the cellular version, the ZymoBIOMICS Microbial Community Standard, so that they can be more powerful when working in tandem.