Learn More
Zymo Research Corporation ZymoBIOMICS™ Microbial Community Standard
SDP
Catalog No. NC1230696
Encompass_Preferred
Microbial composition profiling techniques powered by Next-Generation sequencing are becoming routine in microbiomics and metagenomics studies. However, these analytical techniques can suffer from significant bias from collection to analysis. The ZymoBIOMICS™ Microbial Community Standard is designed to assess bias and errors in the extraction methods of a microbiomics workflow. The Microbial Community Standard mimics a mixed microbial community of well-defined composition, containing three easy-to-lyse Gram-negative bacteria, five tough-to-lyse Gram-positive bacteria and two tough-to-lyse yeasts. Acting as a defined input from the beginning, the Microbial Community Standard can guide construction and optimization of entire workflows and can also be used as a routine quality control.
Catalog No. NC1230696 Supplier Zymo Research Corporation Supplier No. D6300
Microbial composition profiling techniques powered by Next-Generation sequencing are becoming routine in microbiomics and metagenomics studies. However, these analytical techniques can suffer from significant bias from collection to analysis. The ZymoBIOMICS™ Microbial Community Standard is designed to assess bias and errors in the extraction methods of a microbiomics workflow. The Microbial Community Standard mimics a mixed microbial community of well-defined composition, containing three easy-to-lyse Gram-negative bacteria, five tough-to-lyse Gram-positive bacteria and two tough-to-lyse yeasts. Acting as a defined input from the beginning, the Microbial Community Standard can guide construction and optimization of entire workflows and can also be used as a routine quality control.