Learn More
Description
- Less time and less labor work
- ISO and AOAC standards compatible
- No more culture medium preparation
- No extra reagent required
Specifications
Specifications
| Color | Red |
| Detects | Vibrio Parahaemolyticus |
| Form | Prepared Medium |
| Format | Count Plate |
| Product Type | Count Plate |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Identification |
| For Use With (Application) | Microbials Enumeration - Aerobic bacteria, E. coli, Yeast, and Molds etc |
| Incubation Conditions | 36 ±1°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.