Learn More
TRIBIOSCIENCE INC Microbial DNA Magnetic Extraction Kit, 100 Tests
SDP
Catalog No. NC2887471
Encompass_Preferred
Microbial DNA Magnetic Extraction employs magnetic particle-based purification technology to isolate and purify genomic DNA from bacterial and aspergillus contaminated food, plant, herbal, cannabis matrices, and other infused products.
Catalog No. NC2887471 Supplier TRIBIOSCIENCE INC Supplier No. TBS6025100
Microbial DNA Magnetic Extraction employs magnetic particle-based purification technology to isolate and purify genomic DNA from bacterial and aspergillus contaminated food, plant, herbal, cannabis matrices, and other infused products.