Learn More
Sigma Aldrich Fine Chemicals Biosciences Microbial DNA standard fro
Catalog No. 502904318
Encompass
Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification sequencing and bioinformatics analyses. Pseudomonas Aeruginosa genomic DNA microbial standard can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs. Pseudomonas aeruginosa is a gram negative aerobic to facultative anaerobe rod shaped bacteria which commonly inhabits soil and aqueous environments. In cases of a compromised immune system Pseudomonas aeruginosa can be an opportunistic pathogen. Its devastating effect is mostly known in Cystic fibrosis (Cf) patients. in addition it often poses a major problem in burn wounds chronic wounds and in chronic obstructive pulmonary Disorder (COPD).
Catalog No. 50-290-4318 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MBD00140.3UG
Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification sequencing and bioinformatics analyses. Pseudomonas Aeruginosa genomic DNA microbial standard can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs. Pseudomonas aeruginosa is a gram negative aerobic to facultative anaerobe rod shaped bacteria which commonly inhabits soil and aqueous environments. In cases of a compromised immune system Pseudomonas aeruginosa can be an opportunistic pathogen. Its devastating effect is mostly known in Cystic fibrosis (Cf) patients. in addition it often poses a major problem in burn wounds chronic wounds and in chronic obstructive pulmonary Disorder (COPD).