Learn More
Zymo Research Corporation Microbial Selection Buffer, 5 ml
SDP
Catalog No. 501957588
Encompass_Preferred
The Microbial Selection Buffer permits the function of the Microbial Selection Enzyme during Host DNA Depletion in the HostZERO™Microbial DNA Kit.
Catalog No. 50-195-7588 Supplier Zymo Research Corporation Supplier No. D431025
The Microbial Selection Buffer permits the function of the Microbial Selection Enzyme during Host DNA Depletion in the HostZERO™Microbial DNA Kit.