Learn More
Zymo Research Corporation Microbial Selection Enzyme, 50 µl
SDP
Catalog No. 501957589
Encompass_Preferred
The Microbial Selection Enzyme is able to degrade all forms of DNA and RNA and is used during Host DNA Depletion in the HostZERO™Microbial DNA Kit.
Catalog No. 50-195-7589 Supplier Zymo Research Corporation Supplier No. D4310350
The Microbial Selection Enzyme is able to degrade all forms of DNA and RNA and is used during Host DNA Depletion in the HostZERO™Microbial DNA Kit.