Learn More
General Laboratory Products MICROBIO .1PEPTONE 10ML 100/CS
Catalog No. 50821350
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, (0.1%) Peptone Water, 10 ML, 16 x 125 Glass Tube, 100/CS
Catalog No. 50-821-350 Supplier General Laboratory Products Supplier No. 16T240010
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, (0.1%) Peptone Water, 10 ML, 16 x 125 Glass Tube, 100/CS