Learn More
General Laboratory Products MICROBIO 2XLSTW/DT 10ML 100/CS
Catalog No. 50821372
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, (2X) Lauryl Tryptose Broth w/ Durham Tube, 10 ML, 20 x 150 Glass Tube, 100/CS
Catalog No. 50-821-372 Supplier General Laboratory Products Supplier No. 20T1210102X
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, (2X) Lauryl Tryptose Broth w/ Durham Tube, 10 ML, 20 x 150 Glass Tube, 100/CS