Learn More
SPEX CERTIPREP LLC 3P WS - Microbiological PT - 10 Sample Set Part
Catalog No. NC0712720
Encompass_Preferred
(NSI by Zeptometrix) A ten standard set for determination of Total/Fecal Coliforms and E.coli. The standards are designed to be compatible with all promulgated methods including MF,MTF,IDEXX Quanti-Tray,Colilert,and Colisure. With this set, you can report presence/absence and quantitative results. All samples are cultured in the range of 20 - 200 CFU. Sterile hydration buffer included.
Catalog No. NC0712720 Supplier SPEX CERTIPREP LLC 3P Supplier No. MIC001
(NSI by Zeptometrix) A ten standard set for determination of Total/Fecal Coliforms and E.coli. The standards are designed to be compatible with all promulgated methods including MF,MTF,IDEXX Quanti-Tray,Colilert,and Colisure. With this set, you can report presence/absence and quantitative results. All samples are cultured in the range of 20 - 200 CFU. Sterile hydration buffer included.