Learn More
General Laboratory Products MICROBIO TSB 10ML 100/CS
Catalog No. 50821398
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth, 10ML, 20 x 150 Glass Tube, 100/CS
Catalog No. 50-821-398 Supplier General Laboratory Products Supplier No. 20T380010
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth, 10ML, 20 x 150 Glass Tube, 100/CS