Learn More
General Laboratory Products Tryptic Soy Broth (TSB) w/4% Tween 20 & 0.5% Lecithin, 100 mL, 12/cs
Catalog No. NC1812050
Encompass_Preferred
Tryptic Soy Broth w/4% Tween 20 & 0.5% Lecithin -100 ml, 8 oz. wide mouth glass bottle, 12 per case. Base for a General Purpose Medium.
Catalog No. NC1812050 Supplier General Laboratory Products Supplier No. 8WM1180100
Tryptic Soy Broth w/4% Tween 20 & 0.5% Lecithin -100 ml, 8 oz. wide mouth glass bottle, 12 per case. Base for a General Purpose Medium.