Learn More
General Laboratory Products MICROBIO TSBTWLEC 250ML 12/CS
Catalog No. 50821478
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth w/ 4.0% Tween20 and 0.5% Lecithin, 250 ML, W/M Jar, 12/CS
Catalog No. 50-821-478 Supplier General Laboratory Products Supplier No. 16WM1180250
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth w/ 4.0% Tween20 and 0.5% Lecithin, 250 ML, W/M Jar, 12/CS