Learn More
General Laboratory Products MICROBIO TSBTWLEC 490ML 12/CS
Catalog No. 50821509
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth w/ 4.0% Tween20 and 0.5% Lecithin, 490 ML, W/M Jar, 12/CS
Catalog No. 50-821-509 Supplier General Laboratory Products Supplier No. 32WM1180490
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Tryptic Soy Broth w/ 4.0% Tween20 and 0.5% Lecithin, 490 ML, W/M Jar, 12/CS