Learn More
Microbiologics™ Bacteroides fragilis NCTC 9343
SDP
Catalog No. 23016155 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
9343
NCTC 9343
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ATCC®
23-016-155 NCTC 9343
23-013-547 9343
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-016-155 Supplier Microbiologics™ Supplier No. 0358K
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Bacteroides fragilis in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • NCTC 9343
Specifications
ATCC® NCTC 9343
Product Type Quality Control Organism
Species Bacteroides fragilis