Learn More
PHCbi MIR Series 14.3 cu.ft. Large Capacity Microbiological Incubator
Catalog No. 502720048
Encompass_Preferred
The MIR-554-PA large capacity microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.The MIR-554-PA refrigerated incubator provides an ergonomic cabinet design, versatile applications usage and a wide temperature range control.Effective capacity: 14.3 cu.ft. (406 L)Temperature range: -10℃ to +60℃
Catalog No. 50-272-0048 Supplier PHCbi Supplier No. MIR554PA
Add to Cart
Add to Cart
Request a Quote
The MIR-554-PA large capacity microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.The MIR-554-PA refrigerated incubator provides an ergonomic cabinet design, versatile applications usage and a wide temperature range control.Effective capacity: 14.3 cu.ft. (406 L)Temperature range: -10℃ to +60℃