Learn More
Sigma Aldrich Fine Chemicals Biosciences Microbiological monitor po
Catalog No. 502901710
Encompass
features and BenefitsCellulose nitrate membrane with choice of pore sizeSterilized and ready for useEasy handlingEffective time savings up to 70%Black membranes available for better contrastComprehensive range of mediaMonitor workflowMicrobiological monitors have been developed specifically for membrane filter methods for aqueous samples of up to 100 mL. It has never been so easy to identify microorganisms
Catalog No. 50-290-1710 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z69926850EA
features and BenefitsCellulose nitrate membrane with choice of pore sizeSterilized and ready for useEasy handlingEffective time savings up to 70%Black membranes available for better contrastComprehensive range of mediaMonitor workflowMicrobiological monitors have been developed specifically for membrane filter methods for aqueous samples of up to 100 mL. It has never been so easy to identify microorganisms